Learn More
CD68 Monoclonal Antibody (ED1), PE, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA516653
Detalles adicionales : Peso : 0.01000kg
Descripción
Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Mouse anti rat CD68, clone ED1 recognizes the rat ED1 antigen, a heavily glycosylated protein of approximately 90 -110 KDa, also known as rat CD68 (Dijkstra et al. 1985).
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Especificaciones
CD68 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA and 0.09% sodium azide | |
0 | |
Cd68 | |
Rat spleen cells | |
100 Tests | |
Primary | |
Bovine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
ED1 | |
PE | |
Cd68 | |
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
Mouse | |
Protein A | |
RUO | |
287435, 504960 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |