missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Monoclonal Antibody (MEM-111), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MHCD5401
Detalles adicionales : Peso : 0.50000kg
Descripción
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
ICAM-1 Monoclonal antibody specifically detects ICAM-1 in Human samples. It is validated for Flow Cytometry.Especificaciones
ICAM-1, CD54 (ICAM-1) | |
Monoclonal | |
FITC | |
ICAM1 | |
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
Mouse | |
purified | |
ASR, ASR/IVD | |
3383 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
MEM-111 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P05362 | |
ICAM1 | |
Human CD54 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |