missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49c Mouse anti-Human, BUV805, Clone: C3 II.1, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 749378
Detalles adicionales : Peso : 0.00030kg
Descripción
CD49c Monoclonal antibody specifically detects CD49c in Human samples. It is validated for Flow CytometryEspecificaciones
CD49c | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
ITGA3; CD49C; FRP-2; GAP-B3; GAPB3; ILNEB; MSK18; VCA-2; VL3A; VLA3a | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
C3 II.1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 κ |