missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Mouse, FITC, Clone: HI100, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB45-F-25Tests
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD45RA | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
PTPRC | |
Protein A/G Purified | |
RUO | |
5788 | |
IgG2b κ |
Flow Cytometry | |
HI100 | |
FITC | |
NP_002829 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |