missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, anti-Human, FITC, Clone: HI30, STEMCELL Technologies
Mouse Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | CD45 |
---|---|
Clon | HI30 |
Aplicaciones | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
Clasificación | Monoclonal |
Conjugado | FITC |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17137631
|
Stemcell Technologies
100-0466 |
500 Tests |
en la oferta
500 tests |
N/A | Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Este artículo requiere una actualización de precios antes de ser solicitado. Solicite el precio de este artículo aquí. |
|||||||||
Descripción
- The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit II (Catalog #17898; partial blocking may be observed)
Especificaciones
CD45 | |
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
FITC | |
Chimpanzee, Human | |
Primary |
HI30 | |
Monoclonal | |
Mouse | |
IgG1 κ |