missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, NovaFluor™ Blue 530, Clone: SK3 (SK-3), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen H001T02B02

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 17206366

  • 158.00€ / paquete de 25
Fecha estimada de envío: 21-10-2022
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The SK3 monoclonal antibody reacts with human CD4, a 59-kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and at low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.
Applications Reported: This SK3 (SK-3) antibody has been reported for use in flow cytometric analysis.
Applications Tested: This SK3 (SK-3) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 2 μL (0.1 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.
When using more than one NovaFluor dye, CellBlox Monocyte and Macrophage Blocking Buffer may be used to block non-specific binding of dyes to monocytes.
Excitation: 509 nm; Emission: 530 nm; Laser: 488 nm (Blue) Laser

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

CD4
Monoclonal
2 μL/Test
PBS with no preservative; pH 7.2
P01730
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
SK3 (SK-3)
NovaFluor Blue 530
CD4
CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 receptor; CD4, allele 1; cd4a; CD4mut; cell surface glycoprotein CD4; fCD4; L3T4; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); W3/25; W3/25 antigen
Affinity chromatography
RUO
920
4° C, store in dark, DO NOT FREEZE!
Liquid
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales