missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD4 Ab-8, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD4 Ab-8, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1528-S1

Detalles adicionales : Peso : 0.00190kg

 Ver más versiones de este producto

Código de producto. 12699976

  • 555.00€ / 500 microlitros
Fecha estimada de envío: 16-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV.

Host Species: Mouse

Clone: 4B12

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 59 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

CD4 Ab-8
Monoclonal
Unconjugated
Stimulated human leukocytes
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
4B12
Mouse
500 μL
T-Cell
Human
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales