missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse, anti-Human, FITC, Clone: HIT2, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 60014FI.1
Detalles adicionales : Peso : 0.00110kg
Descripción
- The HIT2 antibody reacts with CD38, an ∼45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes
- CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism
- CD31 is a known ligand for the receptor
- In humans, CD38 is expressed by the majority of hematopoietic cells at levels which vary according to the differentiation and activation status of the cells
- It is highly expressed on activated T and B cells and plasma cells, and is also found at high levels in several tissues, including brain, kidney, liver, ovary, pancreas, placenta, testis, and some types of cancerous tissue
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human CD138 Positive Selection Kit II (Catalog #1877) and EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II (Catalog #17887)
Especificaciones
CD38 | |
Monoclonal | |
FITC | |
25 Tests | |
Chimpanzee, Human | |
B Cells, Hematopoietic Stem and Progenitor Cells, Plasma |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
HIT2 | |
Mouse | |
Primary | |
IgG1 κ |