Learn More
CD3 Monoclonal Antibody (17A2), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | CD3 |
---|---|
Clon | 17A2 |
Concentración | 0.2 mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Description: The SK7 monoclonal antibody reacts with human and chimpanzee CD3e, a 20 kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. The SK7 and UCHT1 monoclonal antibodies cross-block binding, suggesting recognition of overlapping epitopes. Applications Reported: This SK7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SK7 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400-42) to minimize any non-specific polymer interactions.
CD3 Monoclonal antibody specifically detects CD3 in Mouse samples. It is validated for Flow Cytometry.Especificaciones
CD3 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
12500, 12501, 12502, 12503 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
17A2 | |
Flow Cytometry | |
Super Bright 702 | |
Rat | |
Mouse | |
IgG2b κ | |
Affinity chromatography | |
Antibody |