missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD275 (B7-H2) Monoclonal Antibody (4H2B1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543376
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human ICOS Ligand/B7-H2/ICOSLG
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.Especificaciones
CD275 (B7-H2) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O75144 | |
ICOSLG | |
Recombinant Human ICOS Ligand/B7-H2/ICOSLG protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
4H2B1 | |
Unconjugated | |
ICOSLG | |
AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
Mouse | |
Protein A | |
RUO | |
23308 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |