missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse, Clone: LPFS2/1611, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 939-MSM2-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD27 | |
Cell Surface | |
LPFS2/1611 | |
Unconjugated | |
XP_016875723 | |
Mouse | |
120kDa | |
Protein A/G Purified | |
RUO | |
939 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD27 | |
Recombinant human CD22 protein | |
Human peripheral blood lymphocytes. Human tonsil and lymph node. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |