missing translation for 'onlineSavingsMsg'
Learn More

CD200 Monoclonal Antibody (OX104), NovaFluor™ Red 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen H038T02R04

103.85 EUR hasta el 2023-03-27
Use el código promocional "20585" para aplicar la oferta.



Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 17860771

  • 155.00€ / paquete de 25
Fecha estimada de envío: 12-04-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The monoclonal antibody OX104 recognizes human CD200 also known as OX2. CD200 is a member of the Ig superfamily with 2 Ig domains, a transmembrane and cytoplasmic domain. CD200 is expressed on resting and activated B cells, a subset of resting and activated T cells, keratinocytes, peripheral and central nerve cells, follicular dendritic cells and ovarian cells. The interaction with CD200R results in macrophage activation (IL-6 production), inhibition of mast cell degranulation along with reduced TNFalpha and IL-13 secretion and overall attenuation of the activation status of lymphocytes. A role has also been suggested in maternal tolerance as expression of CD200 is also present on the trophoblast. Applications Reported: This OX104 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX104 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 0.06μg per test NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Each NovaFluor conjugate or kit is shipped with CellBlox Blocking Buffer. Excitation: 639 nm; Emission: 710 nm; Laser: 633-640 nm (Red) Laser

CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow Cytometry
Especificaciones

Especificaciones

CD200
Monoclonal
0.6 μg/Test
PBS with 0.09% sodium azide; pH 7.2
P41217
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
OX104
NovaFluor Red 710
Cd200
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
Affinity chromatography
RUO
4345
4° C, store in dark, DO NOT FREEZE!
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales