missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 / MS4A1 Rabbit, Clone: IGEL/1497R, enQuireBio™
Rabbit Monoclonal Antibody
Marca: EnquireBio 931-RBM7-P1
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
CD20 / MS4A1 | |
Predominantly cell surface with some cytoplasmic | |
IGEL/1497R | |
Unconjugated | |
NP_690605 | |
Rabbit | |
33-37kDa | |
Protein A/G Purified | |
RUO | |
931 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
MS4A1 | |
Recombinant human full-length MS4A1 protein | |
Daudi, Raji, and U266, and human lymphocytes. Lymph nodes and tonsils. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |