Learn More
CD11c Armenian Hamster, FITC, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marca: EnquireBio QAB23-F-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD11c | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Itgax | |
Protein A/G Purified | |
RUO | |
16411 | |
IgG |
Flow Cytometry | |
N418 | |
FITC | |
NP_067309 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |