Learn More
CD117 (c-Kit) Rat, anti-Mouse, Biotin, Clone: 2B8, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60025BT.1
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17198571
  • 119.70€ / 50 microgramos
Fecha estimada de envío: 14-11-2022
para ver el stock
Descripción
Descripción
  • The 2B8 antibody reacts with CD117 (c-Kit), an ∼145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells
  • CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain
  • Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells
  • The 2B8 antibody does not block binding of c-Kit ligand
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Hematopoietic Progenitor Cell Isolation Kit (Catalog #19856)
Especificaciones
Especificaciones
CD117 (c-Kit)
Monoclonal
Biotin
50 μg
Mouse
Hematopoietic Stem and Progenitor Cells
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry
2B8
Rat
Primary
IgG2b κ