Learn More
CD115 Rat, PE, Clone: AFS98, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB53-PE-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD115 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Csf1r | |
Protein A/G Purified | |
RUO | |
12978 | |
IgG2a κ |
Flow Cytometry | |
AFS98 | |
PE | |
XP_017173299 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |