missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 Monoclonal Antibody (eBioCB-CALLA (CB-CALLA)), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen H054T03Y03
Detalles adicionales : Peso : 0.02000kg
Descripción
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow CytometryEspecificaciones
CD10 | |
Monoclonal | |
0.2 μg/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
P08473 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Flow Cytometry | |
eBioCB-CALLA (CB-CALLA) | |
NovaFluor Yellow 610 | |
Mme | |
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase | |
Affinity chromatography | |
RUO | |
4311 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |