missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT5 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11603-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Especificaciones
CCT5 | |
Polyclonal | |
Unconjugated | |
CCT5 | |
CCT epsilon, CCT5, CCTE, KIAA0098, TCP 1 epsilon | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12465, 22948, 294864, 322258 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48643, P80316, Q68FQ0 | |
cct5 | |
CCT5 Fusion Protein Ag2146 | |
20 μL | |
Primary | |
Human, Zebrafish, Mouse, Rat | |
Antibody | |
IgG |