Learn More
CBP Monoclonal Antibody (NM11), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA513631
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-13631 targets p300/CBP (CREB-Binding Protein) in IF and IHC (P) applications and shows reactivity with Human, Mink, mouse, Non-human primate, and Rat samples. The MA5-13631 immunogen is e1A affinity purified p300 protein.
CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.Especificaciones
CBP | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P45481, Q6JHU9, Q92793 | |
CREBBP | |
E1A affinity purified p300 protein | |
500 μL | |
Primary | |
Human, Mink, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
NM11 | |
Biotin | |
CREBBP | |
AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
Mouse | |
Protein A | |
RUO | |
12914, 1387, 54244 | |
4° C | |
Liquid |