missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C15orf23 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26189-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
C15orf23 is an essential component of the mitotic spindle required for faithful chromosome segregation and progression into anaphase. It promotes the metaphase-to-anaphase transition and is required for chromosome alignment, normal timing of sister chromatid segregation, and maintece of spindle pole architecture. The astrin (SPAG5)-kinastrin (SKAP) complex promotes stable microtubule-kinetochore attachments.Especificaciones
C15orf23 | |
Polyclonal | |
Unconjugated | |
KNSTRN | |
C15orf23, HSD11, KNSTRN, SKAP, TRAF4AF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90417 | |
-20°C | |
Liquid |
Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y448 | |
KNSTRN | |
C15orf23 Fusion Protein Ag21669 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |