missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C12orf32 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5144036
Detalles adicionales : Peso : 0.01000kg
Descripción
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
RHNO1 plays a role in DNA damage response (DDR) signaling upon genotoxic stresses such as ionizing radiation (IR) during the S phase. Recruited to sites of DNA damage through interaction with the 9-1-1 cell-cycle checkpoint response complex and TOPBP1 in a ATR-dependent manner. Required for the progression of the G1 to S phase transition. Plays a role in the stimulation of CHEK1 phosphorylation.Especificaciones
C12orf32 | |
Polyclonal | |
Unconjugated | |
RHNO1 | |
C12orf32; HKMT1188; RAD9, HUS1, RAD1-interacting nuclear orphan protein 1; Rad9, Rad1, Hus1 interacting nuclear orphan; rAD9, RAD1, HUS1-interacting nuclear orphan protein; RAD9-HUS1-RAD1 interacting nuclear orphan 1; RHINO; RHNO1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83695 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Flow Cytometry, Western Blot | |
500 μg/mL | |
PBS with 4MG trehalose and no preservative | |
Q9BSD3 | |
RHNO1 | |
A synthetic peptide corresponding to a sequence of human RHNO1 (RQPSQKAPLLFHQQPLE). | |
100 μg | |
Primary | |
Human | |
Lyophilized |