missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BXDC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17295-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Required for biogenesis of the 60S ribosomal subunit.Especificaciones
BXDC2 | |
Polyclonal | |
Unconjugated | |
BRIX1 | |
BRIX, brix domain containing 2, BRIX1, BXDC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294799, 55299, 67832 | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4QQT6, Q8TDN6, Q9DCA5 | |
BRIX1 | |
BXDC2 Fusion Protein Ag11174 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |