Learn More
BTNL8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558909
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LLRPYIEYPS YNEQNGTPIV ICPVTQESEK EASWQRASAI PETSNSESSS QA Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 32%.
BTNL8 is a single-pass type I membrane protein. It belongs to the immunoglobulin superfamily, BTN/MOG family. BTNL8 contains 1 B30.2/SPRY domain and 1 Ig-like V-type domain. The exact function of BTNL8 remains unknown.Especificaciones
BTNL8 | |
Polyclonal | |
Unconjugated | |
BTNL8 | |
B7-H5 costimulatory molecule; BTN9.2; BTNL8; butyrophilin like 8; butyrophilin-like 8; butyrophilin-like protein 8; UNQ702/PRO1347 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UX41 | |
BTNL8 | |
Recombinant protein corresponding to Human BTNL8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only