missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16330-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency.Especificaciones
BTD | |
Polyclonal | |
Unconjugated | |
BTD | |
Biotinase, biotinidase, BTD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26363, 306262, 686 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43251, Q5FVF9, Q8CIF4 | |
BTD | |
BTD Fusion Protein Ag9449 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |