missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11859-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.Especificaciones
BRD3 | |
Polyclonal | |
Unconjugated | |
BRD3 | |
BRD3, bromodomain containing 3, KIAA0043, ORFX, RING3 like protein, RING3L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362092, 67382, 8019 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15059, Q8K2F0 | |
Brd3 | |
BRD3 Fusion Protein Ag2433 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |