Learn More
Thermo Scientific Chemicals Brain Natriuretic Peptide (1-32), Human
For cellular and molecular biology applications
Marca: Thermo Scientific™ J66167.!D
Detalles adicionales : CAS : 124584-08-3 Peso : 0.00001kg
Descripción
Brain natriuretic peptide is originally isolated from brain, but is mainly produced in myoendocrine cells of the heart ventricles from which it is released into the circulation. It is involved in blood pressure control and cardiovascular homeostasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Human | |
-30°C to -10°C | |
3464.03 | |
0.5 mg | |
BNP (1-32), human |
HPLC | |
Solid | |
Brain Natriuretic Peptide (1-32), human | |
RUO - research use only |
RUO – Research Use Only