missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18933-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).Especificaciones
BMP2 | |
Polyclonal | |
Unconjugated | |
BMP2 | |
BMP 2, BMP 2A, BMP2, BMP2A, bone morphogenetic protein 2, Bone morphogenetic protein 2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
650 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12643 | |
BMP2 | |
BMP2 Fusion Protein Ag13501 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |