Learn More
Uvex™ Blue Type 5/6 Air Disposable Chemical Protection Coveralls
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Descripción
- Exceptional wearer comfort guaranteed by breathable and lightweight SMS material
- Particle-tight and spray-tight to a limited extent
- Ideal for warm working environments and extended periods of wear
- Free of silicone and paint wetting impairment substances
- Material: Polypropylene SMS
- Middle finger loops prevent sleeves sliding up the arm
- Optimal protection in dusty environments
- Self-adhesive zipper flap for optimal protection
- Anti-static properties
- Excellent moisture management
- Optimal fit with elasticated waistband
- Gusset section in crotch area for increased freedom of movement
- Three-part hood for a good fit and unrestricted vision
- Secure and convenient closures thanks to elasticated bands on hood, arms and legs
Especificaciones
Tamaño | X-Large |
Material | Polypropylene |
Color | Blue |
Certificaciones/conformidad | ISO 13982-1 - type 5, EN 13034 - type 6, EN 1149-5, EN 1073-2 |