Learn More
ACROBiosystems Biotinylated Human VSIG8 Protein, Fc, Avitag™
Biotinylated Protein
Marca: ACROBiosystems VS8-H82F2-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
V-set and immunoglobulin domain containing 8 (VSIG8), also known as C1orf204, is a type I transmembrane protein of the B7 family within the Ig superfamily. VSIG8 was identified from proteomic analysis of human hair shafts. It is expressed in the hair follicle and shaft, superficial layers of the nail matrix, and superficial layers of oral epithelium.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 55.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001013683.1 | |
55.5 kDa | |
200 µg | |
Val 22 - Gly 263 | |
VSIG8,C1orf204 | |
Precision Avi | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Biotinylated Human VSIG8, Fc,Avitag (VS8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1). | |
-20°C | |
VSIG8 | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |