missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHMT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15965-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocystinemia, but such a defect has not yet been observed.Especificaciones
BHMT | |
Polyclonal | |
Unconjugated | |
BHMT | |
BHMT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12116, 635 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35490, Q93088 | |
Bhmt | |
BHMT Fusion Protein Ag8738 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |