Learn More
Éster trimetílico de (+/-)-benciloxicarbonil-alfa-fosfonoglicina, 97 %, Thermo Scientific Chemicals
Catalog Number p-7026489
Click to view available options
Cantidad:
1 g
5 g
25 g
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
2-dimetoxifosforil-2-(fenilmetoxicarbonilamino)acetato de metilo
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
2-dimetoxifosforil-2-(fenilmetoxicarbonilamino)acetato de metilo
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El éster trimetílico de (±)-Z-α-fosfonoglicina es el material de partida para la síntesis de (Z)-dehidroaminoácidos por la reacción de Wittig-Horner con aldehídos; los dehidroaminoácidos pueden reducirse estéreoselectivamente a aminoácidos.
Solubilidad
Soluble en metanol.
Notas
Almacenar en un lugar fresco y seco, en recipientes herméticamente cerrados. Almacenar lejos de los agentes oxidantes fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester
Punto de fusión 76°C to 80°C
Cantidad 1 g
Información de solubilidad Soluble in methanol.
Formula Weight (peso de la fórmula) 331.27
Porcentaje de pureza 97%