missing translation for 'onlineSavingsMsg'
Learn More
Learn More
band 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18566-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.Especificaciones
band 3 | |
Polyclonal | |
Unconjugated | |
SLC4A1 | |
AE 1, AE1, Anion exchange protein 1, Anion exchanger 1, band 3, Band 3 anion transport protein, BND3, CD233, DI, EMPB3, EPB3, FR, RTA1A, SLC4A1, SW, WD, WD1, WR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20533, 24779, 6521 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02730, P04919, P23562 | |
SLC4A1 | |
band 3 Fusion Protein Ag13076 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |