missing translation for 'onlineSavingsMsg'
Learn More

band 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

121.38€

Especificaciones

Antígeno band 3
Concentración 0.3 mg/mL
Aplicaciones Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16818544
Ver documentos Detalles de la promoción
Proteintech
18566-1-AP-20UL
20 μL Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A Ver productos alternativos
Ahorre hasta 
»
Descripción

Descripción

The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.
Especificaciones

Especificaciones

band 3
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
P02730, P04919, P23562
20533, 24779, 6521
band 3 Fusion Protein Ag13076
Primary
-20°C
SLC4A1
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AE 1, AE1, Anion exchange protein 1, Anion exchanger 1, band 3, Band 3 anion transport protein, BND3, CD233, DI, EMPB3, EPB3, FR, RTA1A, SLC4A1, SW, WD, WD1, WR
SLC4A1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications