missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ HDPE weighing scoop
Light weight and chemical resistant
Marca: Azlon FWC320
Detalles adicionales : Peso : 0.01100kg
Descripción
- Lightweight HDPE weighing boats
- Chemical resistant
Especificaciones
2 mL | |
Boat | |
33 x 15 mm | |
15 mm |
HDPE | |
Weighing Scoop | |
40 mm |