Learn More
SI Analytics™ Automatic 3/2 Way Valve
SI Analytics™ Automatic 3/2 Way Valve is for use with wide range of products including interchangable modules.
Marca: SI Analytics™ TZ3000
Detalles adicionales : Peso : 0.01000kg
Descripción
- For interchangeable modules TA 05 plus through TA 50 plus and WA 05 through WA 50
- Suitable for TitroLine™ Easy and 5000, TitroLine™ KF, TITRONIC™ Universal, Basic, and 300
Especificaciones
Automatic 3-way Valve | |
TitroLine Alpha Plus, TitroLine KF, and TITRONIC 110 Plus |