missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B (Proliferation Marker) Mouse, Clone: AURKB/1593, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 9212-MSM3-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Aurora B (Proliferation Marker) | |
Nuclear and Cytoplasmic | |
AURKB/1593 | |
Unconjugated | |
Mouse | |
39kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
9212 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Aurora B protein fragment (aa89-251) | |
HeLa cells. Tonsil, Placenta, Hodgkin's Lymphoma or Colon Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |