missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17589-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Especificaciones
ATP5H | |
Polyclonal | |
Unconjugated | |
ATP5H | |
ATP5H, ATP5JD, ATPase subunit d, ATPQ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10476, 641434, 71679 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75947, P31399, Q9DCX2 | |
Atp5h | |
ATP5H Fusion Protein Ag11429 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |