missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11946-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.Especificaciones
ATF1 | |
Polyclonal | |
Unconjugated | |
ATF1 | |
ATF1, EWS ATF1, FUS/ATF 1, Protein TREB36, TREB36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11908, 315305, 466 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18846, P81269 | |
Atf1 | |
ATF1 Fusion Protein Ag2548 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |