missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14116-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ASPH is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins. This gene is thought to play an important role in calcium homeostasis. Alternative splicing of this gene results in five transcript variants which vary in protein translation, the coding of catalytic domains, and tissue expression. Variation among these transcripts impacts their functions which involve roles in the calcium storage and release process in the endoplasmic and sarcoplasmic reticulum as well as hydroxylation of aspartic acid and asparagine in epidermal growth factor-like domains of various proteins.Especificaciones
ASPH | |
Polyclonal | |
Unconjugated | |
ASPH | |
ASP beta hydroxylase, aspartate beta hydroxylase, ASPH, BAH, CASQ2BP1, HAAH, JCTN, junctin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
444 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12797 | |
ASPH | |
ASPH Fusion Protein Ag5265 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |