missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10618-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. Alternative splicing results in multiple transcript variants.Especificaciones
AP1M2 | |
Polyclonal | |
Unconjugated | |
AP1M2 | |
AP 1 complex subunit mu 2, AP mu chain family member mu1B, AP1 mu2, AP1M2, HSMU1B, MU 1B, Mu adaptin 2, MU1B, Mu1B adaptin, mu2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10053, 11768, 367038 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9WVP1, Q9Y6Q5 | |
Ap1m2 | |
AP1M2 Fusion Protein Ag0961 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |