missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Trace Amine Receptor 1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24720
Detalles adicionales : Peso : 0.00970kg
Descripción
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Especificaciones
Trace Amine Receptor 1 | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
Rabbit | |
Protein G purified | |
RUO | |
134864 | |
Human, Primate | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Q96RJ0 | |
TAAR1 | |
A portion of amino acids 225-250 of human TAAR1 was used as the immunogen | |
0.1 mg | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% homologous in mouse and 80% homologous in dog. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
For Research Use Only