Learn More
SFRS10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541788
Additional Details : Peso : 0.00010kg
Description
Peptide sequence: GVFGLSLYTT ERDLREVFSK YGPIADVSIV YDQQSRRSRG FAFVYFENVD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%
SFRS10 Polyclonal antibody specifically detects SFRS10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SFRS10 | |
Polyclonal | |
Unconjugated | |
TRA2B | |
5730405G21Rik; D16Ertd266e; Htra2-beta; PPP1R156; protein phosphatase 1, regulatory subunit 156; RA301; serine/arginine-rich splicing factor 10; SFRS10; SIG-41; Silg41; Silica-induced gene 41 protein; similar to splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich 10; splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich, 10; SRFS10; Srsf10; TRA-2 beta; Tra2b; TRA2beta; TRA2-beta; TRAN2B; transformer 2 beta homolog; transformer 2 beta homolog (Drosophila); transformer-2 protein homolog B; Transformer-2 protein homolog beta; zgc:56612 | |
Rabbit | |
Protein A | |
RUO | |
6434 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P62995 | |
TRA2B | |
Synthetic peptide directed towards the middle region of human SFRS10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |