Learn More
PKN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534784
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (86%), Rat (86%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKN2 Polyclonal antibody specifically detects PKN2 in Human samples. It is validated for Western Blot.Especificaciones
PKN2 | |
Polyclonal | |
Unconjugated | |
PKN2 | |
6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; p140 kinase; PAK2; Pak-2; PKN gamma; PKN2; PRK2; PRKCL2; PRO2042; protease-activated kinase 2; protein kinase C-like 2; protein kinase N2; protein-kinase C-related kinase 2; RP5-905H16.1; serine/threonine kinase 7; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2; serine/threonine-protein kinase N2-like; si:ch211-83f14.1; Stk7; wu:fa97c07 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5586 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q16513 | |
PKN2 | |
Recombinant fragment contains a sequence corresponding to a region within amino acids 487 and 984 of PKN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |