Learn More
PI3K p85 alpha Monoclonal Antibody (U13), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA121472
Detalles adicionales : Peso : 0.01000kg
Descripción
MA1-21472 detects PI 3 Kinase p85 alpha in Human, Non-human primate, Mouse and Rat samples. MA1-21472 has been successfully used in Immunoprecipitation and Western Blot procedures. MA1-21472 immunogen corresponds to Baculovirus expressed (recombit) Bovine p85 alpha subunit of PI 3 Kinase..
Phosphatidylinositol 3-kinase (PIK3) is a signaling protein that phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. Signaling is initiated when upstream tyrosine kinase receptors or G protein coupled receptors are bound by their ligands. It binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance. During insulin stimulation, it also binds to IRS-1. Mutations in the gene can result in Agammaglobulinemia 7, SHORT syndrome, or Immunodeficiency 36.Especificaciones
PI3K p85 alpha | |
Monoclonal | |
Unconjugated | |
P27986, P26450, Q63787 | |
PIK3R1 | |
Baculovirus expressed (recombit) Bovine p85 alpha subunit of PI 3 Kinase. | |
RUO | |
18708, 25513, 5295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
U13 | |
PIK3R1 | |
GRB1, p85, p85-ALPHA | |
Mouse | |
1 mL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |