missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ADD1/ADD2 (Ser726, Ser713) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536614
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 81 kDa. Purity is >95% by SDS-PAGE.
Phospho-ADD1/ADD2 (Ser726, Ser713) Polyclonal antibody specifically detects Phospho-ADD1/ADD2 (Ser726, Ser713) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-ADD1/ADD2 (Ser726, Ser713) | |
Polyclonal | |
Unconjugated | |
ADD1 | |
2900072M03Rik; ADD1; Add2; Add97; ADDA; ADDB; adducin 1; adducin 1 (alpha); adducin 2; adducin 2 (beta); adducin beta; Adducin1; Adducin-1; adducin-63; Adducin-alpha; Adducin-beta; AI256389; alpha Adducin; alpha-ADD; alpha-adducin; beta Adducin; beta-ADD; beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha; Erythrocyte adducin subunit beta; snoRNA MBII-396 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11518, 11519, 118, 119, 24170, 24171 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35611, P35612, Q05764, Q63028, Q9QYB8, Q9QYC0 | |
ADD1, ADD2 | |
Synthetic phosphopeptide derived from human Adducin alpha around the phosphorylation site of Serine 726 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |