Learn More
K-cadherin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529984
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: H1299, HCT116. Predicted reactivity: Mouse (97%), Rat (98%), Chicken (85%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
K-cadherin Polyclonal antibody specifically detects K-cadherin in Human samples. It is validated for Western Blot.Especificaciones
K-cadherin | |
Polyclonal | |
Unconjugated | |
CDH6 | |
CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P55285 | |
CDH6 | |
Recombinant fragment corresponding to a region within amino acids 325 and 606 of Human K cadherin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |