missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgM κ, Rat, R-PE, Clone: R4-22, Isotype Control, BD
Marca: BD Biosciences 553943
Detalles adicionales : Peso : 0.01000kg
Descripción
The R4-22 antibody has unknown specificity.
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Cytotoxicity Assay, Isotype Control
Especificaciones
Cytotoxicity Assay | |
0.2mg/mL | |
Rat | |
0.1 mg | |
IgM κ |
R4-22 | |
R-PE | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |