Learn More
GST-π Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610719
Detalles adicionales : Peso : 0.01000kg
Descripción
Glutathione-S-Transferases (GSTs) are a family of dimeric proteins that catalyze the S-conjugation of glutathione with many compounds for detoxification. The GSTs are categorized into four classes based on their biochemical and structural properties: α, μ, π, and θ. GST-π is of particular interest because it is over-expressed in many tumors, but is either absent or expressed at low levels in the corresponding normal tissues. This high expression of GST-π is associated with malignant transformation and correlates with a decrease in GST activity. Three different GST-π proteins have been isolated and designated as hGSTP1™A, hGSTP1™B, and hGSTP1™C. These isoforms are nearly identical, varying by only one to two amino acids.
Immunofluorescence, Immunoprecipitation, Western Blotting
Especificaciones
GST-π | |
Monoclonal | |
250μg/mL | |
Glutathione-S-Transferase-π | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
3 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human GST-π aa. 5-210 | |
150 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG1 |
For Research Use Only.