Learn More
gamma Tubulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534815
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, HeLa, Jurkat, Raji, NCI-H929, NIH3T3, rat kidney. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (98%), Xenopus laevis (99%), Dog (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.Especificaciones
gamma Tubulin | |
Polyclonal | |
Unconjugated | |
TUBG1 | |
1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103733, 103768, 252921, 27175, 393882, 680991, 7283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
0.95 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P23258, P83887, P83888, Q8VCK3, Q9NRH3 | |
TUBG1, Tubg2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 184 of gamma Tubulin (Uniprot ID#P23258) | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |