missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALR2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518630
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
GALR2 Polyclonal antibody specifically detects GALR2 in Human samples. It is validated for Western Blot.Especificaciones
GALR2 | |
Polyclonal | |
Unconjugated | |
Galr2 | |
Gal receptor 2; Gal-2 receptor; GAL2-R; Galanin r2 receptor; galanin receptor 2; galanin receptor type 2; Galanin-2 receptor; Galnr2; GALR2; Gal-r2; GALR-2; mGalR; MGC125983; MGC125984 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8811 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43603 | |
Galr2 | |
Synthetic peptide sequence (RYPLHSRELRTPRN) corresponding to the internal amino acids of GALR2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |